Learn More
Socorex Positive Displacement Micropipetter Replacement Plungers from DWK Life Sciences™
Shop All DWK Life Sciences Products

Description
- PTFE Tipped
- Draws Sample Into Precision Glass Capillary Tube
Specifications
Specifications
| Color | Green |
| Material | Stainless Steel |
| Type | Replacement Plunger |
| Volume (Metric) | 10 to 50 μL |
| For Use With (Equipment) | Socorex Acura Positive Displacement Micropipetter |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.