Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Socorex Positive Displacement Micropipetter Replacement Plungers from DWK Life Sciences™
SDP

Catalog No. 1370755 Shop All DWK Life Sciences Products
Change view
Click to view available options
Volume (Metric):
10 to 50 μL
100 to 200 μL
Color:
Green
Red
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Color
13-707-55 100 to 200 μL Red
13-707-53 10 to 50 μL Green
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 13-707-55 Supplier Socorex Supplier No. 851334
Add to Cart
Add to Cart

100 - 200μL Plungers for Acura 841 Positive Displacement Micropipette

  • PTFE Tipped
  • Draws Sample Into Precision Glass Capillary Tube
TRUSTED_SUSTAINABILITY

Specifications

Color Red
Material Stainless Steel
Type Replacement Plunger
Volume (Metric) 100 to 200 μL
For Use With (Equipment) Socorex Acura Positive Displacement Micropipetter
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.