Learn More
Description
- Genuine Socorex spare parts for Dosys self-refilling syringes
- Spare grounded glass syringe barrels available in multiple capacities
- Lab cannula available for dispensing applications
- Designed to maintain the precision and performance of Dosys syringes
- Supplied by DWK Life Sciences, guaranteeing authentic Socorex components
Specifications
Specifications
| Material | Silicone |
| For Use With (Application) | Liquid handling |
| Length (English) | 0.1181 in. |
| Length (Metric) | 0.299 cm |
| Product Type | Spare Barrel |
| For Use With (Equipment) | Self-refilling Syringe |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
