Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences Wheaton™ Spin Vanes for V-Vials
GSA_VASDP

Catalog No. p-3303345 Shop All DWK Life Sciences Products
Change view
Click to view available options
For Use With (Equipment):
Fit 0.3 and 1mL V-Vials
Fit 2, 3, 5, and 10mL V-Vials
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
22-021824 Fit 0.3 and 1mL V-Vials
22-021825 Fit 2, 3, 5, and 10mL V-Vials
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 22-021824 Supplier DWK Life Sciences Supplier No. 903061
Only null left
Add to Cart
Add to Cart

Magnetic stir bar for use with NextGen™ V Vials™

  • Made from PTFE
  • Place stir bar in vial and use with magnetic stirrer

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Material PTFE
Shape Triangle
Quantity 6
For Use With (Equipment) Fit 0.3 and 1mL V-Vials
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.