Learn More
DWK Life Sciences Wheaton™ Square Media Bottles with Standard Caps
SDP
Catalog No. 02924974 Shop All DWK Life Sciences Products
Change view
Click to view available options
Capacity (Metric):
250 mL
1000 mL
2000 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
02-924-974 250 mL
02-924-977 1000 mL
02-924-979 2000 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 02-924-974 Supplier DWK Life Sciences Supplier No. WPBGC0250SB
Add to Cart
Add to Cart
This item is not returnable. View return policy
Square/Octagonal Media Bottles made of PETG that dispalys clarity, toughness, chemical inertness and acts as gas barrier
  • For storage of media, laboratory reagents and biological buffers
  • Made of Eastman Tritan Eastar Copolyester PETG that is resin used for single use labware and medical devices
  • With standard blue polypropylene caps
  • Molded graduation markings for reference filling
  • Gamma irradiated sterilized to SAL 10-6
  • Cleanroom packaged for sterile production with multi-barrier elastomer packaging
  • Resin meets ISO 10993 and/or USP Class VI biocompatibility requirements and is Food Contact Status compliant
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 250 mL
Material PETG