Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences Wheaton™ Stainless-Steel Shaft Assembly Kit For Celstir
SDP

Catalog No. 02911580 Shop All DWK Life Sciences Products
Change view
Click to view available options
For Use With (Equipment):
1000mL Flask
125mL Flask
250mL Flask
6000mL Flask
8000mL Flask
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
02-911-580 8000mL Flask
02-911-577 250mL Flask
02-911-576 125mL Flask
02-911-578 1000mL Flask
02-911-579 6000mL Flask
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 02-911-580 Supplier DWK Life Sciences Supplier No. 356891
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Adjustable paddle blade impeller for better mixing

  • Provide maximum surface interface between culture and flask atmosphere
  • Ideal for microcarrier and suspension cultures such as insect cells, hybridomas, and adapted cell lines
  • Made from Wheaton 33 low-extractable borosilicate glass that conforms to ASTM™ Type I, Class A and USP Type I requirements
  • Impeller rod does not protrude up through top cap
  • Sidearms for 500mL to 8L flask allow vented caps which enables the flask to breathe while protecting from contamination
  • 1:1 headspace for good gas exchange
  • No contamination from stirring rod
  • Bottom dimple on 125mL and larger flask improves circulation and reduces accumulation of cells in center of the flask
  • Sizes 125 to 1000mL are graduated in 50mL increments; sizes 3000 to 8000mL are graduated in 500mL increments; sizes 25mL and 50mL are non-graduated
  • Completely autoclavable at 121°C for 20 minutes

Order Info

Assemblies 500mL and larger can be used with the vented cap (Cat. No. 22-288-526)

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Equipment) 8000mL Flask
Type Shaft Assembly Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.