Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DWK Life Sciences Wheaton™ Stainless-Steel Shaft Assembly Kit For Celstir

Click to view available options
For Use With (Equipment):
1000mL Flask
125mL Flask
250mL Flask
6000mL Flask
8000mL Flask
Description
- Provide maximum surface interface between culture and flask atmosphere
- Ideal for microcarrier and suspension cultures such as insect cells, hybridomas, and adapted cell lines
- Made from Wheaton 33 low-extractable borosilicate glass that conforms to ASTM™ Type I, Class A and USP Type I requirements
- Impeller rod does not protrude up through top cap
- Sidearms for 500mL to 8L flask allow vented caps which enables the flask to breathe while protecting from contamination
- 1:1 headspace for good gas exchange
- No contamination from stirring rod
- Bottom dimple on 125mL and larger flask improves circulation and reduces accumulation of cells in center of the flask
- Sizes 125 to 1000mL are graduated in 50mL increments; sizes 3000 to 8000mL are graduated in 500mL increments; sizes 25mL and 50mL are non-graduated
- Completely autoclavable at 121°C for 20 minutes
Order Info
Assemblies 500mL and larger can be used with the vented cap (Cat. No. 22-288-526)

Specifications
Specifications
For Use With (Equipment) | 250mL Flask |
Type | Shaft Assembly Kit |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction