Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences WHEATON™ Stainless Steel Support Screens
SDP

Catalog No. p-3303932 Shop All DWK Life Sciences Products
Change view
Click to view available options
For Use With (Equipment):
25 mm Vacuum Filtration Assembly
47mm Vacuum Filtration Assembly
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
14-209-120 47mm Vacuum Filtration Assembly
14-209-107 25 mm Vacuum Filtration Assembly
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 14-209-120 Supplier DWK Life Sciences Supplier No. 419364
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For vacuum filtration assembly. WHEATON™ Stainless Steel Support Screens are manufactured from the highest quality materials.

For vacuum filtration assembly. Wheaton™ Stainless Steel Support Screens are manufactured from the highest quality materials.

  • Made from stainless steel for durability and chemical resistance
  • Designed for use with vacuum filtration assemblies
  • Compatible with standard 25mm and 47mm filtration assemblies
TRUSTED_SUSTAINABILITY

Specifications

Disposable Reusable
Material Stainless Steel
Sterility Non-sterile
For Use With (Equipment) 47mm Vacuum Filtration Assembly
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.