Learn More
DWK Life Sciences WHEATON™ Black Phenolic Screw Cap with Open Top
Shop All DWK Life Sciences Products

Description
- Autoclavable black phenolic screw cap for reliable, repeatable sterilisation
- Open top design allows syringe access without cap removal
- Linerless construction eliminates liner contamination risk
- Compatible with standard screw-thread laboratory bottles and vials
Specifications
Specifications
| Color | Black |
| Material | Phenolic Resin |
| Sterility | Non-sterile |
| Liner Material | N/A |
| For Use With (Equipment) | Vacule Ampules |
| Closure Size | 15-425 |
| Autoclavable | Autoclavable |
| Disposable | Reusable |
| Product Type | Screw Cap |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.