Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences WHEATON™ Stoppers for Serum Bottles and Vials: Locking Flange Septa
SDP

Catalog No. 0645059 Shop All DWK Life Sciences Products
Change view
Click to view available options
Color:
Gray
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
06-450-59 Gray
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 06-450-59 Supplier DWK Life Sciences Supplier No. 240563
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

WHEATON™ Stoppers for Serum Bottles and Vials: Locking Flange Septa provide airtight covers for serum bottles and vials. Flange-type stoppers are secured with aluminum crimp seals; slotted and three-prong plug stoppers are designed for freeze-dry applications.

Provide airtight covers

  • Flange-type stoppers are secured with aluminum crimp seals
  • Slotted and three-prong plug stoppers are for freeze-dry applications
TRUSTED_SUSTAINABILITY

Specifications

Diameter (Metric) Bottom 20.8 mm
Diameter (Metric) Outer 33 mm
Diameter (Metric) Top 33 mm
Product Type Straight Plug Stopper
Shape Rounded
Sterility Non-sterile
Autoclavable Autoclavable
Diameter (Metric) 3.3 cm
Material Chlorobutyl Rubber
For Use With (Equipment) Serum bottles and vials
Color Gray
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.