Learn More
DWK Life Sciences WHEATON™ Thin Flange Stopper for Serum Bottles and Vials
Shop All DWK Life Sciences Products

Description
- Flange-type stoppers are secured with aluminum crimp seals
- Slotted and three-prong plug stoppers are ideal for freeze-dry applications
- Autoclavable
- Shrink-wrapped partitioned packaging
Specifications
Specifications
| Diameter (Metric) Bottom | 13 mm |
| Diameter (Metric) Outer | 20 mm |
| Diameter (Metric) Top | 20 mm |
| Shape | Cylindrical |
| Autoclavable | Autoclavable |
| Quantity | 300 Stoppers/Pk. |
| Description | 3-leg |
| For Use With (Equipment) | Serum Bottles and Vials 1.3 cm I.D. and 2 cm O.D. |
| Sterility | Non-sterile |
| Product Type | Thin Flange Stopper |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.