Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences Wheaton™ Stoppers for Serum Bottles and Vials: Straight Plug Stopper
GSA_VASDP

Catalog No. 0640611D Shop All DWK Life Sciences Products
Change view
Click to view available options
Color:
Gray
Red
Material:
Butyl Rubber
Isoprene Rubber
Rubber
Diameter (Metric):
13 mm
30 mm
20 cm
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Material Diameter (Metric)
06-406-11D Gray Isoprene Rubber 13 mm
06-447J Gray Butyl Rubber 20 cm
06-451-61 Red Rubber 30 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 06-406-11D Supplier DWK Life Sciences Supplier No. 224100072
Only null left
Add to Cart
Add to Cart

Provide airtight covers

  • Flange-type stoppers are secured with aluminum crimp seals
  • Slotted and three-prong plug stoppers are for freeze-dry applications

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Equipment) Serum bottles and vials
Diameter (Metric) Inner 7 mm
Autoclavable Autoclavable
Diameter (Metric) Bottom 7 mm
Product Type Straight Plug Stopper
Diameter (Metric) 13 mm
Material Isoprene Rubber
Hardness 40 Durometer
Color Gray
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.