Learn More
DWK Life Sciences Wheaton™ Straight Plug Style Silicone Stoppers for Vials and Serum Bottles
SDP
Catalog No. 03084138 Shop All DWK Life Sciences Products
Change view
Click to view available options
Material:
Silicone
Catalog No. Material
03-084-138 Silicone
1 options
Catalog No. 03-084-138 Supplier DWK Life Sciences Supplier No. 224100178
Add to Cart
Add to Cart
For maximum sealing of vials with straight wall glass finish
  • Available in choice of different materials (silicone and bromobutyl)
Not recommended for vials with a blow-back feature.
TRUSTED_SUSTAINABILITY
Specifications
Autoclavable Autoclavable
Diameter (Metric) Inner 2 cm
Product Type Straight Plug Stopper
Hardness 55 Durometer
Material Silicone
For Use With (Equipment) Fits 1.3cm I.D., 2cm O.D. mouth of vials and serum bottles