Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences WHEATON™ Thin Flange Stopper for Serum Bottles and Vials
SDP

Catalog No. 01549775 Shop All DWK Life Sciences Products
Change view
Click to view available options
Stopper Number:
20
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Stopper Number
01-549-775 20
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-549-775 Supplier DWK Life Sciences Supplier No. W224100190
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

WHEATON™ Thin Flange Stopper for Serum Bottles and Vials provides airtight covers for serum bottles and vials. Secured with aluminum crimp seals or used as snap-on closures, these autoclavable stoppers are compatible with screw neck diagnostic bottles.

  • Flange-type stoppers are secured with aluminum crimp seals
  • Snap-on design offers convenient closure for screw neck diagnostic bottles
  • Compatible with WHEATON screw neck diagnostic bottles and I-Loc™ closures
  • Autoclavable for repeated sterilization use
  • Available in 20mm size with gray chlorobutyl construction
TRUSTED_SUSTAINABILITY

Specifications

Diameter (Metric) Bottom 13 mm
Diameter (Metric) Outer 20 mm
Diameter (Metric) Top 20 mm
Product Type Thin Flange Stopper
Description Snap-on
For Use With (Equipment) Serum Bottles and Vials 1.3 cm I.D. and 2 cm O.D.
Shape Ribbed Cone
For Use With (Application) Lyophilization
Color Gray
Sterility Non-sterile
Material Chlorobutyl Rubber
Stopper Number 20
Quantity 300 Stoppers/Pk.
Diameter (Metric) Inner 2 cm
Autoclavable Autoclavable
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.