Learn More
DWK Life Sciences Wheaton™ Tissue Grinder Kit
SDP
Catalog No. 0841417 Shop All DWK Life Sciences Products
Change view
Click to view available options
Product Type:
Micro tissue grinder kit
Catalog No. Product Type
08-414-17 Micro tissue grinder kit
1 options
Catalog No. 08-414-17 Supplier DWK Life Sciences Supplier No. 358204
Add to Cart
Add to Cart
Includes
1mL Tenbroeck; 0.5mL grinder with 13-425 GPI cap; 0.1 and 0.2mL standard; 2mL Potter-Elvehjem; 1mL tapered
Complete selection of micro tissue grinders packed in a protective carrying case
Autoclavable
Order Info
Replacement parts available separately
TRUSTED_SUSTAINABILITY
Specifications
Product Type Micro tissue grinder kit
Includes 1mL Tenbroeck, 0.5mL grinder with 13 to 425mm GPI cap, 0.1 and 0.2mL standard, 2mL Potter-Elvehjem, 1mL tapered