Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences™ Wheaton™ Tubular Serum Vials
SDP

Catalog No. 01670189 Shop All DWK Life Sciences Products
Change view
Click to view available options
Volume (Metric):
4 mL
10 mL
13.5 mL
Diameter (Metric) Outer:
13 mm
22 mm
24 mm
Height (Metric):
35 mm
40 mm
45 mm
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Diameter (Metric) Outer Height (Metric)
01-670-189 13.5 mL 24 mm 45 mm
01-670-187 4 mL 13 mm 35 mm
01-670-188 10 mL 22 mm 40 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01-670-189 Supplier DWK Life Sciences™ Supplier No. WEBB10RTEP
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Tubular Serum Vials are certified ISO 8362-1.

  • Clear vials are converted from Type I borosilicate glass tubing
  • Amber vials converted from borosilicate glass tubing that conforms to USP Type I requirements for light transmission
TRUSTED_SUSTAINABILITY

Specifications

Color Clear
Diameter (Metric) Outer 24 mm
Material Borosilicate Glass
Pre-slit No
Sterility Non-sterile
Disposable Single-use
Volume (Metric) 13.5 mL
Graduated No
Includes Vial
Quantity 150 Vials/Tray
Marking Spot No
For Use With (Equipment) Stopper, Cap
Autoclavable Autoclavable
Height (Metric) 45 mm
Product Type Serum Vial
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.