Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences Wheaton™ White Polypropylene Caps with Foam/LDPE Liners
SDP

Catalog No. 06450103 Shop All DWK Life Sciences Products
Change view
Click to view available options
Neck Finish:
15-415
20-410
24-400
24-410
28-400
28-410
33-400
33-430
Liner Material:
Foam/LDPE
Polyethylene Foam
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Neck Finish Liner Material
06-450-103 24-400 Polyethylene Foam
06-450-97 15-415 Polyethylene Foam
06-450-105 28-400 Foam/LDPE
06-450-101 20-410 Polyethylene Foam
06-450-104 24-410 Foam/LDPE
06-450-107 33-400 Foam/LDPE
06-450-106 28-410 Foam/LDPE
06-450-108 33-430 Foam/LDPE
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 06-450-103 Supplier DWK Life Sciences Supplier No. 242220
Add to Cart
Add to Cart

For general storage and packaging applications

  • Lined with foam polyethylene
  • Excellent barrier to aqueous solutions
  • Repeat sealability

TRUSTED_SUSTAINABILITY

Specifications

Color White
Material Polypropylene
No. per Case 200 Cs.
Liner Material Polyethylene Foam
Neck Finish 24-400
Certifications/Compliance ISO 9001
For Use With (Equipment) Glass bottle or vial
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.