Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences Wheaton™ White Polypropylene Caps with Foamed Polyvinyl Liners
SDP

Catalog No. 0298438 Shop All DWK Life Sciences Products
Change view
Click to view available options
Neck Finish:
13-425
24-400
28-400
38-400
43-400
48-400
53-400
58-400
70-400
Material:
Glass
Polypropylene
No. per Case:
144 Cs.
48 Cs.
72 Cs.
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Neck Finish Material No. per Case
02-984-38 43-400 Polypropylene 72 Cs.
02-984-50 58-400 Polypropylene 72 Cs.
02-984-44 48-400 Polypropylene 72 Cs.
02-984-4 13-425 Polypropylene 144 Cs.
02-984-47 53-400 Glass 72 Cs.
02-984-24 24-400 Polypropylene 144 Cs.
02-984-56 70-400 Polypropylene 48 Cs.
02-984-28 28-400 Polypropylene 144 Cs.
02-984-35 38-400 Polypropylene 72 Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. 02-984-38 Supplier DWK Life Sciences Supplier No. 239286
Add to Cart
Add to Cart

Foamed polyethylene liner provides chemical resistance for acids, alkalis, solvents, alcohols, oils, household cosmetics and aqueous products

  • Liner provides tight seal
  • Liner Material: Foam, Polyethylene
  • Polypropylene
  • Screw Cap
  • White

Use these caps with glass bottles or vials

Order Info

Each pack contains 72

TRUSTED_SUSTAINABILITY

Specifications

Color White
Material Polypropylene
No. per Case 72 Cs.
Neck Finish 43-400
Certifications/Compliance ISO 9001
For Use With (Equipment) Glass bottle or vial
Closure Liner Polyethylene Foam
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.