Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences WHEATON™ White Polypropylene Caps with Foamed Polyethylene Liners
SDP

Catalog No. 0298456 Shop All DWK Life Sciences Products
Change view
Click to view available options
Neck Finish:
13-425
24-400
28-400
38-400
43-400
48-400
53-400
58-400
70-400
Material:
Polypropylene
No. per Case:
144 Cs.
48 Cs.
72 Cs.
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Neck Finish Material No. per Case
02-984-56 70-400 Polypropylene 48 Cs.
02-984-38 43-400 Polypropylene 72 Cs.
02-984-50 58-400 Polypropylene 72 Cs.
02-984-44 48-400 Polypropylene 72 Cs.
02-984-4 13-425 Polypropylene 144 Cs.
02-984-47 53-400 Polypropylene 72 Cs.
02-984-24 24-400 Polypropylene 144 Cs.
02-984-28 28-400 Polypropylene 144 Cs.
02-984-35 38-400 Polypropylene 72 Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. 02-984-56 Supplier DWK Life Sciences Supplier No. 239292
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

WHEATON™ White Polypropylene Caps with Foamed Polyethylene Liners deliver a reliable tight seal with broad chemical resistance against acids, alkalis, solvents, alcohols, oils, and aqueous products. Compatible with glass bottles and vials.

  • Foamed polyethylene liner provides a tight, secure seal
  • Chemical resistance for acids, alkalis, solvents, alcohols, oils, and aqueous products
  • White polypropylene screw cap construction
  • Compatible with glass bottles and vials
  • Available in multiple cap sizes to suit a range of bottle and vial formats

Order Info

Each pack contains 72

TRUSTED_SUSTAINABILITY

Specifications

Color White
Material Polypropylene
No. per Case 48 Cs.
Sterility Non-sterile
Neck Finish 70-400
For Use With (Equipment) Glass bottle or vial
Closure Liner Polyethylene Foam
Autoclavable Not Autoclavable
Certifications/Compliance ISO 9001
Disposable Reusable
Product Type Cap
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.