Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences Wheaton™ White Polypropylene Caps with PTFE-Faced Foamed Polyethylene Liners
GSA_VASDP

Catalog No. 0298430 Shop All DWK Life Sciences Products
Change view
Click to view available options
Neck Finish:
20-400
22-400
24-400
28-400
33-400
38-400
43-400
45-400
48-400
53-400
58-400
70-400
89-400
Liner Material:
PTFE/Polyethylene
Polyethylene Foam
Material:
Polypropylene
Polypropylene, PTFE
No. per Case:
144 Cs.
48 Cs.
72 Cs.
13 product options available for selection
Product selection table with 13 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Neck Finish Liner Material Material No. per Case
02-984-30 33-400 PTFE/Polyethylene Polypropylene 144 Cs.
02-984-14 20-400 PTFE/Polyethylene Polypropylene 144 Cs.
02-984-26 28-400 PTFE/Polyethylene Polypropylene 144 Cs.
02-984-46 53-400 PTFE/Polyethylene Polypropylene 72 Cs.
02-984-22 24-400 PTFE/Polyethylene Polypropylene, PTFE 144 Cs.
02-984-37 43-400 PTFE/Polyethylene Polypropylene 72 Cs.
02-984-40 45-400 Polyethylene Foam Polypropylene 72 Cs.
02-984-18 22-400 Polyethylene Foam Polypropylene 144 Cs.
02-984-43 48-400 PTFE/Polyethylene Polypropylene 72 Cs.
02-984-49 58-400 PTFE/Polyethylene Polypropylene 72 Cs.
02-984-34 38-400 PTFE/Polyethylene Polypropylene 72 Cs.
02-984-61 89-400 PTFE/Polyethylene Polypropylene 48 Cs.
02-984-55 70-400 PTFE/Polyethylene Polypropylene 48 Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 13 options available.
13 options
Catalog No. 02-984-30 Supplier DWK Life Sciences Supplier No. 239236
Only null left
Add to Cart
Add to Cart

Excellent for environmental samples, pharmaceuticals and diagnostic reagents

  • PTFE faced foamed polyethylene liner offers chemical resistance
  • Polyethylene foam provides compressibility and sealing properties
  • Good for high purity chemicals, strong acids, and solvents
  • For usewith glass bottles or vials
  • Polypropylene
  • Screw cap
  • White

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Color White
Material Polypropylene
No. per Case 144 Cs.
Liner Material PTFE/Polyethylene
Neck Finish 33-400
Certifications/Compliance ISO 9001
For Use With (Equipment) Glass bottle or vial
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.