Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences WHEATON™ White Polypropylene Screw Caps for Glass Bottles and Vials
SDP

Catalog No. 03084122 Shop All DWK Life Sciences Products
Change view
Click to view available options
Neck Finish:
15-425
18-400
24-400
Liner Material:
PTFE/Polyethylene
PTFE/Silicone
Polyethylene Foam
Polyvinyl
No. per Case:
144/Cs.
200/Cs.
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Neck Finish Liner Material No. per Case
03-084-122 18-400 Polyethylene Foam 144/Cs.
03-084-113 24-400 PTFE/Silicone 200/Cs.
03-084-119 15-425 Polyethylene Foam 144/Cs.
03-084-121 18-400 Polyvinyl 144/Cs.
03-084-123 18-400 PTFE/Polyethylene 144/Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 03-084-122 Supplier DWK Life Sciences Supplier No. 239275
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

WHEATON™ White Polypropylene Screw Caps for Glass Bottles and Vials provide a reliable, chemical-resistant closure with a clear surface for labelling. Available in open or closed top styles with multiple liner options to suit a wide range of laboratory applications.

  • White polypropylene construction with clear surface for writing or labelling
  • Available in open top or closed top configurations
  • Multiple liner options to suit different application requirements
  • Caps with PTFE/silicone liner: provides heat resistance, glass-filled construction, suited for EPA samples
  • Caps with foamed polyethylene liner: chemical resistant, delivers a tight seal
  • Caps with poly-vinyl liner: moisture barrier, suited for mild acids, alkalis, solvents, alcohols, oils and aqueous products
  • Caps with PTFE faced foamed polyethylene liner: chemical resistant, provides compressibility and sealing properties, suited for high purity chemicals, strong acids and solvents
TRUSTED_SUSTAINABILITY

Specifications

Color White
Material Polypropylene
No. per Case 144/Cs.
Sterility Non-sterile
Liner Material Polyethylene Foam
Neck Finish 18-400
For Use With (Equipment) Glass bottle or vial
Autoclavable Not Autoclavable
Certifications/Compliance ISO 9001
Disposable Reusable
Product Type Screw Cap
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.