Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences Wheaton™ White Polypropylene Screw Caps for Glass Bottles and Vials
SDP

Catalog No. p-4608864 Shop All DWK Life Sciences Products
Change view
Click to view available options
Neck Finish:
15-425
18-400
24-400
Liner Material:
PTFE/Polyethylene
PTFE/Silicone
Polyethylene Foam
Polyvinyl
No. per Case:
144/Cs.
200/Cs.
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Neck Finish Liner Material No. per Case
03-084-113 24-400 PTFE/Silicone 200/Cs.
03-084-119 15-425 Polyethylene Foam 144/Cs.
03-084-122 18-400 Polyethylene Foam 144/Cs.
03-084-121 18-400 Polyvinyl 144/Cs.
03-084-123 18-400 PTFE/Polyethylene 144/Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 03-084-113 Supplier DWK Life Sciences Supplier No. W224600
Only null left
Add to Cart
Add to Cart

With clear surface for writing

  • Available with different liners (PTFE/silicone, foamed polyethylene, poly-vinyl, PTFE faced foamed polyethylene)
  • With choice of open or closed top

Caps with PTFE/silicone liner:

  • Provides heat resistance
  • Glass filled
  • Suited for EPA samples
Caps with foamed polyethylene liner:
  • Chemical resistant
  • Tight seal
Caps with poly-vinyl liner:
  • With moisture barrier
  • Suited for mild acids, alkalis, solvents, alcohols, oils and aqueous products
Caps with PTFE faced foamed polyethylene liner:
  • Chemical resistant
  • Provides compressibility and sealing properties
  • Suited for high purity chemicals, strong acids and solvents

TRUSTED_SUSTAINABILITY

Specifications

Color White
Material Polypropylene
No. per Case 200/Cs.
Liner Material PTFE/Silicone
Neck Finish 24-400
Certifications/Compliance ISO 9001
For Use With (Equipment) Glass EPA vial
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.