Learn More
Sigma Aldrich Fine Chemicals Biosciences WHEATONR amber sample vial
Catalog No. 502891542
Encompass
Amber Sample Vials in Lab File (With Caps Attached). Attached caps help maintain cleanliness. Supplied in partitioned trays to provide an easy way to store empty vials or to inventory samples.Amber vials are manufactured from Wheaton 320 glass that conforms to USP Type I requirements to protect light-sensitive samples. Screw caps are attached to vials. It is suggested to use rubber lined caps for aqueous samples, and PTFE faced rubber lined caps for organic samples. Vials and caps are autoclavable. 2 mL vials fit the M-T Vial File, 228778. 4 mL vials fit the M-T Vial File, 228780.
Catalog No. 50-289-1542 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. DWKW22460472EA
Amber Sample Vials in Lab File (With Caps Attached). Attached caps help maintain cleanliness. Supplied in partitioned trays to provide an easy way to store empty vials or to inventory samples.Amber vials are manufactured from Wheaton 320 glass that conforms to USP Type I requirements to protect light-sensitive samples. Screw caps are attached to vials. It is suggested to use rubber lined caps for aqueous samples, and PTFE faced rubber lined caps for organic samples. Vials and caps are autoclavable. 2 mL vials fit the M-T Vial File, 228778. 4 mL vials fit the M-T Vial File, 228780.