Learn More
Sigma Aldrich Fine Chemicals Biosciences WHEATONR CELLine Multiuse
Supplier:  Sigma Aldrich Fine Chemicals Biosciences DWKWCL1000AD33EA
Encompass
CELLine AD Bioreactor Flask for high density adherent cell culture.Media compartment with cell growth compartment for antibody and protein production.Use with Hybridoma Insect CHO NSO HEK BHK cell linesDisposable and ready-to-useHigh call density and high product concentrationBulk media chamber allows for longer run times without media refreshing and reduces operation timeDecreases use of consumablesCost-efficient space saving and stackableNo additional equipment required for operation
Catalog No. 50-290-8707