Learn More
Sigma Aldrich Fine Chemicals Biosciences WHEATONR clear sample vial
Supplier:  Sigma Aldrich Fine Chemicals Biosciences DWKW224582144EA
Encompass
Sample Vials in Lab File (With Caps Attached). Attached caps help maintain cleanliness. Supplied in partitioned trays to provide an easy way to store empty vials or to inventory samples.Screw caps are attached to vials. It is suggested to use rubber lined caps for aqueous samples, and PTFE faced rubber lined caps for organic samples, and metal foil liner for hydrocarbons. Vials and caps are autoclavable. 2 mL vials fit the M-T Vial File, 228778. 4 mL vials fit the M-T Vial File, 228780.Vials with caps attached packaged in partitioned trays
Catalog No. 50-289-1554