Learn More
Sigma Aldrich Fine Chemicals Biosciences WHEATONR CryoELITER cryovi
Catalog No. 502904291
Encompass
CryoELITE cryogenic vial with Loctagon skirt provides stability in freestanding position and locks into the CryoELITE Benchmate Rack making it easy to open and close the vial with one hand. The unique manufacturing design of the CryoELITE vial creates more cap to vial surface contact for an absolute seal. The vial bottom format provides unrestricted sample viewing. This results in exceptional cap seal performance exceeding DOT standards by an average of 35% making the CryoELITE vial ideal for the transport of clinical samples. Caps and vials are made from low binding cryogenic virgin polypropylene. Unrivaled cap seal exceeds DOT and IATA regulations ensuring ultimate protection of samples during transportation and demanding freeze-thaw handlingMade from low binding cryogenic grade virgin polyproplene Lot certified RNase/DNase and Endotoxin free providing assurance of product integrityInternal threaded screw cap can be easily removed with one handSterile.nb
Catalog No. 50-290-4291 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. DWKW985865500EA
CryoELITE cryogenic vial with Loctagon skirt provides stability in freestanding position and locks into the CryoELITE Benchmate Rack making it easy to open and close the vial with one hand. The unique manufacturing design of the CryoELITE vial creates more cap to vial surface contact for an absolute seal. The vial bottom format provides unrestricted sample viewing. This results in exceptional cap seal performance exceeding DOT standards by an average of 35% making the CryoELITE vial ideal for the transport of clinical samples. Caps and vials are made from low binding cryogenic virgin polypropylene. Unrivaled cap seal exceeds DOT and IATA regulations ensuring ultimate protection of samples during transportation and demanding freeze-thaw handlingMade from low binding cryogenic grade virgin polyproplene Lot certified RNase/DNase and Endotoxin free providing assurance of product integrityInternal threaded screw cap can be easily removed with one handSterile.nb