Learn More
Sigma Aldrich Fine Chemicals Biosciences WHEATONR CryoELITER wide m
Catalog No. 502904444
Encompass
CryoELITE Tissue Vial was developed in response to demand from researchers looking for a secure reliable cryogenically approved vial to store and transport cryogenic tissues. featuring the proven Wheaton unrivaled cap seal that exceeds DOT and IATA regulations the Wheaton CryoELITE Tissue vial allows consumers to leverage Wheatons expertise in cryogenic storage for cells and fluids to tissues. Shipped sterile in cases containing 10 bags of 25 vials with certificates of sterility and RNase/DNase and Endotoxin free the Wheaton CryoELITE Tissue vial is the newest innovative SOLUTIONon designed by Wheaton to support the research community.Wide-mouth CryoELITE vial for use with tissue sections Unrivaled cap seal exceeds DOT and IATA regulations ensuring ultimate protection of samples during transportation and demanding freeze-thaw handlingMade from low-binding cryogenic grade virgin polypropyleneLot certified RNase/DNase and endotoxin free and nonpyrogenic providing
Catalog No. 50-290-4444 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. DWKW985100250EA
CryoELITE Tissue Vial was developed in response to demand from researchers looking for a secure reliable cryogenically approved vial to store and transport cryogenic tissues. featuring the proven Wheaton unrivaled cap seal that exceeds DOT and IATA regulations the Wheaton CryoELITE Tissue vial allows consumers to leverage Wheatons expertise in cryogenic storage for cells and fluids to tissues. Shipped sterile in cases containing 10 bags of 25 vials with certificates of sterility and RNase/DNase and Endotoxin free the Wheaton CryoELITE Tissue vial is the newest innovative SOLUTIONon designed by Wheaton to support the research community.Wide-mouth CryoELITE vial for use with tissue sections Unrivaled cap seal exceeds DOT and IATA regulations ensuring ultimate protection of samples during transportation and demanding freeze-thaw handlingMade from low-binding cryogenic grade virgin polypropyleneLot certified RNase/DNase and endotoxin free and nonpyrogenic providing