Learn More
Sigma Aldrich Fine Chemicals Biosciences WHEATONR liquid scintillat
Catalog No. 502904025
Encompass
Glass Liquid Scintillation Vials with separately packs caps20 mL sizeMade from Wheaton 180 low potassium borosilicate glassBackground counts are consistent and low ultraviolet transmission is highDimensions (OD x H) 28 mm x 61 mm (with cap attached)
Catalog No. 50-290-4025 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. DWK986580500EA
Glass Liquid Scintillation Vials with separately packs caps20 mL sizeMade from Wheaton 180 low potassium borosilicate glassBackground counts are consistent and low ultraviolet transmission is highDimensions (OD x H) 28 mm x 61 mm (with cap attached)