Learn More
Sigma Aldrich Fine Chemicals Biosciences WHEATONR liquid scintillat
Supplier:  Sigma Aldrich Fine Chemicals Biosciences DWK986706500EA
Encompass
Liquid Scintillation Vials with caps attachedThese 20 mL vials are manufactured from high density polyethylene. Vials with caps attached are packaged in individual cells in trays of 100. Vials with screw caps packed separately are packaged in polyethylene bags of 100 and placed in a separate tray. Cases of 1000 vials are packed in a single plastic bag with caps in a separate bag. Trays of 100 vials are shrink-wrapped in plastic for protection against contamination.20 mL sizeMade from high density polyethyleneAvailable with screw cap attached or with caps packed separatelyDimensions (OD x H) 27 mm x 61 mm (with cap attached)Shrink-wrapped in plastic
Catalog No. 50-290-4623