Learn More
Sigma Aldrich Fine Chemicals Biosciences WHEATONR liquid scintillat
Supplier: Sigma Aldrich Fine Chemicals Biosciences DWK986754500EA
Liquid Scintillation Vials with separately packs capsfor optimal performance in the laboratory PET plastic vials provide the clarity of glass and have the safety of using plastic. Vials can be safely incinerated no harmful gas is generated only CO2 and H2O are created. Lightweight and virtually unbreakable PET vials offer low permeability to solvents and minimal background counts. 20 mL size Made from polyethylene terephthalate (PET)Clarity of glass with safety of plastic Low permeability to solvents and minimal background countDimensions (OD x H) 27 mm x 61 mm (with cap attached)