Learn More
Sigma Aldrich Fine Chemicals Biosciences WHEATONR liquid scintillat
Supplier:  Sigma Aldrich Fine Chemicals Biosciences DWK986561500EA
Encompass
Glass Liquid Scintillation Vials with separately packs caps20 mL sizeMade from Wheaton 180 low potassium borosilicate glassBackground counts are consistent and low ultraviolet transmission is highAvailable with cap attached caps packed separately or without capDimensions (OD x H) 28 mm x 61 mm (with cap attached)
Catalog No. 50-290-4775