Learn More
Sigma Aldrich Fine Chemicals Biosciences WHEATONR serum vial tube c
Catalog No. 502900464
Encompass
Serum tubing vials. Glass can withstand the challenges of reduced pressure and extreme temperature variations. Protects against changes in pH and maintain the purity of the contentsClear vials manufactured from low extractable borosilicate glass that conforms to USP type 1 and ASTM E 438 Type 1 Class A requirementsAmber glass conforms to USP 1 requirements to protect light-sensitive samplesTubular design provides excellent clarity and dimensional consistency from vial to vialSpecially designed bottom radius adds strength for lyophilization applicationsLighter weight compared to molded bottlesShrink-wrapped modules reduce particulate contaminationAutoclavable
Catalog No. 50-290-0464 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. DWK223685144EA
Serum tubing vials. Glass can withstand the challenges of reduced pressure and extreme temperature variations. Protects against changes in pH and maintain the purity of the contentsClear vials manufactured from low extractable borosilicate glass that conforms to USP type 1 and ASTM E 438 Type 1 Class A requirementsAmber glass conforms to USP 1 requirements to protect light-sensitive samplesTubular design provides excellent clarity and dimensional consistency from vial to vialSpecially designed bottom radius adds strength for lyophilization applicationsLighter weight compared to molded bottlesShrink-wrapped modules reduce particulate contaminationAutoclavable