Learn More
Sigma Aldrich Fine Chemicals Biosciences WHEATONR Shorty Vials clea
Catalog No. 502904679
Encompass
20 mL Glass scintillation vials with cttached caps suitable for strong solvents and high-purity chemicals. Glass 20 mL scintillation vials with attached caps. Chemically inert suitable for strong solvents and high-purity chemicalsVials packaged in cellular trays which have alphanumeric coordinates for fast easy identification.27 mm OD x 57 mm heightCases shrink-wrapped to preserve cleanlinessVials are manufactured from 33 expansion low extractable borosilicate glass conforming to ASTM E438 Type I Class A requirements
Catalog No. 50-290-4679 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. DWKW224608200EA
20 mL Glass scintillation vials with cttached caps suitable for strong solvents and high-purity chemicals. Glass 20 mL scintillation vials with attached caps. Chemically inert suitable for strong solvents and high-purity chemicalsVials packaged in cellular trays which have alphanumeric coordinates for fast easy identification.27 mm OD x 57 mm heightCases shrink-wrapped to preserve cleanlinessVials are manufactured from 33 expansion low extractable borosilicate glass conforming to ASTM E438 Type I Class A requirements