Learn More
Grainger Zoro Select Aluminum Wheel Chock, Grooved, 7 in x 8 in x 11-5/8 in, 1 ea
Catalog No. 502778953
Encompass_Preferred
This single, general purpose wheel chock is engineered from durable aluminum to provide reliable stabilization for commercial vehicles. Featuring a grooved surface tread, this chock is designed to help maintain vehicle positioning.
  • Constructed from aluminum for robust performance.
  • Grooved surface tread design to enhance grip.
  • Single wheel chock style.
  • Includes a sign with the text 'CHOCK YOUR WHEELS' and an integrated flag for high visibility.
  • Dimensions measure 7 in. L x 8 in. H x 11 5/8 in. W.
Catalog No. 50-277-8953 Supplier Grainger Supplier No. 19P544
This single, general purpose wheel chock is engineered from durable aluminum to provide reliable stabilization for commercial vehicles. Featuring a grooved surface tread, this chock is designed to help maintain vehicle positioning.
  • Constructed from aluminum for robust performance.
  • Grooved surface tread design to enhance grip.
  • Single wheel chock style.
  • Includes a sign with the text 'CHOCK YOUR WHEELS' and an integrated flag for high visibility.
  • Dimensions measure 7 in. L x 8 in. H x 11 5/8 in. W.