Learn More
Grainger Approved Vendor Wheel Chock, Urethane, 9-1/4 in Length, 1 ea
Catalog No. 502782807
Encompass_Preferred
These urethane wheel chocks provide a lightweight and durable solution for securing commercial vehicles. Constructed from materials resistant to chemicals, fuel, oil, and harsh weather, they offer a reliable alternative to traditional steel or iron chocks, facilitating easier handling during placement and removal.
  • Constructed from urethane material for durability
  • Chemical, fuel, oil, and weather resistant
  • Grooved surface and base tread for enhanced grip
  • Lightweight design for ease of use
  • Suitable for commercial vehicle applications
Catalog No. 50-278-2807 Supplier Grainger Supplier No. 19P545
These urethane wheel chocks provide a lightweight and durable solution for securing commercial vehicles. Constructed from materials resistant to chemicals, fuel, oil, and harsh weather, they offer a reliable alternative to traditional steel or iron chocks, facilitating easier handling during placement and removal.
  • Constructed from urethane material for durability
  • Chemical, fuel, oil, and weather resistant
  • Grooved surface and base tread for enhanced grip
  • Lightweight design for ease of use
  • Suitable for commercial vehicle applications