Learn More
Alkali Scientific Wheel Controller / Filler, green, 10mL
SDP
Catalog No. 5000662927
Encompass_Preferred
Economical manual pipette pump designed for accurate and controlled liquid dispensing in laboratory applications. The unit features a smooth thumbwheel mechanism that allows precise filling and dispensing of liquids while maintaining consistent control during use. A secure tapered collar holds both glass and plastic pipettes firmly in place to prevent slipping during operation. Liquids can be quickly dispensed by pressing the white side lever or plunger, enabling fast and efficient workflow. The green color coding supports easy volume identification for 10 mL capacity. The pump disassembles easily for convenient cleaning and maintenance, ensuring long service life and reliable performance. Its ergonomic design allows comfortable one handed operation, making it ideal for routine laboratory liquid handling tasks.
Catalog No. 50-006-62927 Supplier Alkali Scientific Supplier No. P7491
Economical manual pipette pump designed for accurate and controlled liquid dispensing in laboratory applications. The unit features a smooth thumbwheel mechanism that allows precise filling and dispensing of liquids while maintaining consistent control during use. A secure tapered collar holds both glass and plastic pipettes firmly in place to prevent slipping during operation. Liquids can be quickly dispensed by pressing the white side lever or plunger, enabling fast and efficient workflow. The green color coding supports easy volume identification for 10 mL capacity. The pump disassembles easily for convenient cleaning and maintenance, ensuring long service life and reliable performance. Its ergonomic design allows comfortable one handed operation, making it ideal for routine laboratory liquid handling tasks.