Learn More
Medicus Health Wheeled 45 Quart Blood Bank Transport Cooler
SDP
Catalog No. NC3310716
Encompass
Maxi-Cool Rotomolded Wheeled 45 Quart Blood Bank Transport Cooler comes with a convenient telescopic handle to make moving large volumes of specimens easy ∙ 45 Quart Wheeled Blood Bank Transport Cooler goes beyond the minimum standard days of ice retention, It can store up to 10 days or longer  if pre-cooled, kept in low temps & UV index  ∙ Cooler is food grade safe, dry ice compatible and tough enough to endure whatever the medical environment throws your way ∙ Cooler labeled with "Return to Blood Bank" clearly blasted on the front∙ Smooth surfaces are easy to clean and disinfect with no retention of bio-burden∙ Super duty nylon rope handles (removable)∙ Dry ice compatible∙ External Dimensions: 22”W x 18”D x 16”H; Internal Dimensions: 16.5”W x 11.5”D x 14”H
Catalog No. NC3310716 Supplier Medicus Health Supplier No. 5305M3
Maxi-Cool Rotomolded Wheeled 45 Quart Blood Bank Transport Cooler comes with a convenient telescopic handle to make moving large volumes of specimens easy ∙ 45 Quart Wheeled Blood Bank Transport Cooler goes beyond the minimum standard days of ice retention, It can store up to 10 days or longer  if pre-cooled, kept in low temps & UV index  ∙ Cooler is food grade safe, dry ice compatible and tough enough to endure whatever the medical environment throws your way ∙ Cooler labeled with "Return to Blood Bank" clearly blasted on the front∙ Smooth surfaces are easy to clean and disinfect with no retention of bio-burden∙ Super duty nylon rope handles (removable)∙ Dry ice compatible∙ External Dimensions: 22”W x 18”D x 16”H; Internal Dimensions: 16.5”W x 11.5”D x 14”H