Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aqua Phoenix™ Where's the Water Deluxe Kit Components
SCI_ED

Catalog No. S396325WW Shop All Fisherbrand Products
Change view
Click to view available options
Product Type:
Beaker
Medicine cups
Mini plastic animals
Mixing bowl
Tacky putty
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
S396325WW Beaker
S396385WW Mixing bowl
S396395WW Medicine cups
S396415WW Tacky putty
S396435WW Mini plastic animals
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. S396325WW
Add to Cart
Add to Cart

Replacement parts for Educational Kit

Variety of components specially formatted as replacements for AquaPhoenix custom designed hands-on educational kits.

Specifications

Grade K to 5
For Use With (Application) Education Kit
Material HDPE
Capacity (Metric) 1000 mL
Product Type Beaker
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.