Learn More
Abcam WHI-P131 hydrochloride, JAK3 inhibitor, 25MG
Catalog No. 502706322
Encompass_Preferred
MW 297.31 Da, Purity >98%. Cell-permeable Janus kinase 3 (JAK3) inhibitor (IC₅₀ values of 9.1 μM and 78 μM for human and mouse recombinant proteins respectively). Selective versus JAK1, JAK2, SYK, BTK and LYN tyrosine kinases.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6322 Supplier Abcam Supplier No. AB14212625MG
MW 297.31 Da, Purity >98%. Cell-permeable Janus kinase 3 (JAK3) inhibitor (IC₅₀ values of 9.1 μM and 78 μM for human and mouse recombinant proteins respectively). Selective versus JAK1, JAK2, SYK, BTK and LYN tyrosine kinases.
The product is subject to the following: Abcam Restricted Use Statement