Learn More
Medchemexpress LLC HY-108419 5mg Medchemexpress, WHI-P258 CAS:21561-09-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1084195MG
Encompass_Preferred
Medchemexpress, HY-108419 5mg WHI-P258 CAS:21561-09-1 WHI-P258, a quinazoline compound, binds to the active site of JAK3 with an estimated Ki of 72 µM. WHI-P258 does not inhibit JAK3 and does not affect the thrombin-induced aggregation of platelets even at 100 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8545