Learn More
Grainger Whip 48 in L Silver Handle PK2
Supplier:  Grainger 14D395
Encompass_Preferred
The 14D395 is a 14-inch diameter, 395 CFM, direct drive exhaust fan made by Dayton. It features a thermally protected PSC motor with permanently lubricated ball bearings for maintenance-free operation. The fan has a galvanized steel housing and is designed for wall mounting with the included mounting bracket. At the maximum speed of 1075 RPM, this exhaust fan produces a sound level of 68 dBA. The 14D395 operates on 115V power and is ETL listed. With proper installation, this Dayton exhaust fan provides high airflow for ventilation in commercial and industrial settings.
Catalog No. 50-275-3455