Learn More
Weber Scientific Whirl-Pak Sterile Sampling Bags, 58 mL, write-on strip, 500/pk
SDP
Catalog No. NC1410562
Encompass
Sterile, single service bags are made of heavy transparent polyethylene. Leak-proof and airtight. Puncture-proof tabs. Capacity is measured when the bag is closed and tab is folded over 3 times. Bags should not be used at temperatures above 180°F (82°C). Bags can be frozen to any temperature. Write-on strips have a paper-like and smear-resistant finish and can be written on with an ordinary ballpoint pen, pencil or marker.
Catalog No. NC1410562 Supplier Weber Scientific Supplier No. 303200
Sterile, single service bags are made of heavy transparent polyethylene. Leak-proof and airtight. Puncture-proof tabs. Capacity is measured when the bag is closed and tab is folded over 3 times. Bags should not be used at temperatures above 180°F (82°C). Bags can be frozen to any temperature. Write-on strips have a paper-like and smear-resistant finish and can be written on with an ordinary ballpoint pen, pencil or marker.