Learn More
Sigma Aldrich Fine Chemicals Biosciences Whirl-PakR Sample Bag 710
Catalog No. 502900394
Encompass
Capacity is measured in fluid ounces when the bag is closed and tab is folded over three times. Volume and dimensions are approximate bags should not be used at temperatures above 180 f (82 C). Bags can be frozen to any temperature even liquid nitrogen temperatures down to -346 f (-210 C) - careful handling required after freezing. All bags are sterilized after manufacturing.
Catalog No. 50-290-0394 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. WPB01020WA500EA
Capacity is measured in fluid ounces when the bag is closed and tab is folded over three times. Volume and dimensions are approximate bags should not be used at temperatures above 180 f (82 C). Bags can be frozen to any temperature even liquid nitrogen temperatures down to -346 f (-210 C) - careful handling required after freezing. All bags are sterilized after manufacturing.