Learn More
Sigma Aldrich Fine Chemicals Biosciences Whirl-PakR Stand-Up Bag 40
Catalog No. 502905465
Encompass
The Stand-Up Bag makes sample handling and testing much easier because it gives users the ability to perform important laboratory functions with both hands because the bag will stand completely by itself. No rack or other holder is needed to keep the bag upright. When the sample is placed inside the special gusseted bottom flattens out allowing the bag to stand up on its own. It works just like a beaker or bottle however it costs less is sterilized wont break and uses only a fraction of storage space as the other containers.The white write-on strip makes sample identification easier and more convenient. Its easy to write on this white strip using an ordinary ballpoint pen pencil or felt-tip marker. The paper-like finish virtually eliminates ink smearing and provides a surface that will accept a variety of writing instruments.
Catalog No. 50-290-5465 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. WPB01542WA100EA
The Stand-Up Bag makes sample handling and testing much easier because it gives users the ability to perform important laboratory functions with both hands because the bag will stand completely by itself. No rack or other holder is needed to keep the bag upright. When the sample is placed inside the special gusseted bottom flattens out allowing the bag to stand up on its own. It works just like a beaker or bottle however it costs less is sterilized wont break and uses only a fraction of storage space as the other containers.The white write-on strip makes sample identification easier and more convenient. Its easy to write on this white strip using an ordinary ballpoint pen pencil or felt-tip marker. The paper-like finish virtually eliminates ink smearing and provides a surface that will accept a variety of writing instruments.