Learn More
Sigma Aldrich Fine Chemicals Biosciences Whirl-PakR Write-On Bag 27
Catalog No. 502900457
Encompass
The white write-on strip makes sample identification easier and more convenient. Its easy to write on this white strip using an ordinary ballpoint pen pencil or felt-tip marker. The paper-like finish virtually eliminates ink smearing and provides a surface that will accept a variety of writing instruments.
Catalog No. 50-290-0457 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. WPB01445WA250EA
The white write-on strip makes sample identification easier and more convenient. Its easy to write on this white strip using an ordinary ballpoint pen pencil or felt-tip marker. The paper-like finish virtually eliminates ink smearing and provides a surface that will accept a variety of writing instruments.