Learn More
International Enviroguard™ White Anti-skid Shoe Cover with Black Vinyl Sole
Catalog No. 17184151 Shop All International Enviroguard Systems Products
Change view
Click to view available options
Closure Type:
Elastic Top
Catalog No. Closure Type
17-184-151 Elastic Top
1 options
Catalog No. 17-184-151 Supplier International Enviroguard™ Supplier No. 4100
Add to Cart
Add to Cart
Anti-skid shoe covers use proprietary fabric that helps filter out hazardous particulates as small as 0.3 microns in the 95 to 99% range and resist abrasions and use breathable fabric to keep the feet cool.
  • Shoe covers resist abrasions and use breathable fabric to reduce heat buildup
  • Secure elastic closures help keep shoe covers in place, while an anti-skid sole provides better traction to avoid slips, trips, and falls
  • Vinyl soles help improve traction to reduce slips, trips, and falls
  • A high moisture vapor transmission rate (MVTR) allows heat and moisture vapor to escape so wearers stay cooler
Specifications
Size Universal
Material Microporous Laminate
Latex Free Yes
Color White
Disposable Single-use
Sterility Non-sterile
Closure Type Elastic Top
Sole Type Anti-skid
Resistance Airborne Solid Particulates, Dry Particulates, Abrasion, Liquid Aerosol
Product Type Shoe Covers