Learn More
International Enviroguard™ White Biohazard Coveralls with Attached Boots
Catalog No. 17183960 Shop All International Enviroguard Systems Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-183-960 2X-Large
17-183-955 Small
17-183-956 Medium
17-183-957 Large
17-183-954 X-Large
17-183-959 3X-Large
17-183-958 4X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 17-183-960 Supplier International Enviroguard™ Supplier No. W25012XL
Add to Cart
Add to Cart
ViroGuard 2 disposable biohazard suits are designed with fabric and seams that have been rigorously tested for protection against blood and bloodborne pathogens.
  • Durable fabric is built to withstand tears and abrasions in tough working environments
  • Both fabric and seams pass ASTM F1670 and F1671 tests for protection against blood, bodily fluids, and bloodborne pathogens
  • Zipper front with a sealable storm flap helps keep contaminants from entering or seeping through otherwise-exposed zipper seams
  • Reinforced with double-sided tape at the wrists, your gloves will remain secure as you work
  • Red fabric pull tab on the zipper makes it quick and easy to find so you can suit up or down in no time
  • Taped seams seal off seams from contaminants, while elastic wrists and ankles keep coveralls secure and in place to avoid skin exposure
  • For added convenience, thumb and finger loops are included to help prevent sleeves from riding up
  • Attached boots protect your feet while latex-free, elastic wrists offer a secure fit to prevent skin exposure of the arms
  • Mandarin collar with hook and loop closure at neck provides a better neck seal
  • Roomy, one-piece raglan sleeves resist tears around shoulders
  • Gusseted crotch gives the user more freedom to move, bend, crouch, climb, and squat without ripping fabric
  • Possible applications include emergency response, healthcare, category 3 black water, sanitation work, crime scene remediation, and animal research
Specifications
Product Type A50 Breathable Particle and Splash Protection Coveralls
Size 2X-Large
Gender Unisex
Material Multi-layer Barrier Laminate Fabric
Color White
Disposable Single-use
Waist Style Elastic at Back
Ankle Style Attached Boots
Cuff Style Elastic, Finger Loops
Neck Style Collared
Hood Type Standard
Closure Type 2-way Zipper with Sealed Storm Flap
Seam Type Serged, Taped
Sterility Non-sterile
Resistance Bloodborne Pathogens, Liquid, Particle, Tears
Latex Free Yes
No. of Pockets None