Learn More
International Enviroguard™ White Biohazard Hood with Taped Seams
Catalog No. 17183944 Shop All International Enviroguard Systems Products
Change view
Click to view available options
Color:
White
Catalog No. Color
17-183-944 White
1 options
Catalog No. 17-183-944 Supplier International Enviroguard™ Supplier No. W2561
Add to Cart
Add to Cart
ViroGuard 2 hoods with hook and loop closure are designed with fabric and taped seams that have been rigorously tested for protection against blood and bloodborne pathogens.
  • Durable fabric is built to withstand tears and abrasions in tough working environments
  • Taped seams add an extra layer of protection, helping to prevent contaminants from entering through exposed seams
  • Seams are also impermeable to blood, bodily fluids, and bloodborne pathogens
  • Face opening uses latex-free elastic and the hood is secured using an easy hook and loop closure
  • Hood length provides protection of the head, neck, and shoulders for more coverage of the upper body
  • Impermeable fabric and seams pass ASTM F1670 and F1671 tests for protection against blood, bodily fluids, and bloodborne pathogens
  • Latex-free elastic face opening
  • Possible applications include emergency response, healthcare, category 3 black water, crime scene remediation, animal research, and more
Specifications
Product Type Overhood
Size Universal
Material Laminate
Color White
Latex Free Yes
Includes Taped Seams