Learn More
International Enviroguard™ White Biohazard Suit with Elastic Wrists and Ankles
Catalog No. 17183948 Shop All International Enviroguard Systems Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-183-948 Small
17-183-949 Medium
17-183-950 Large
17-183-947 X-Large
17-183-953 2X-Large
17-183-952 3X-Large
17-183-951 4X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 17-183-948 Supplier International Enviroguard™ Supplier No. W2502S
Add to Cart
Add to Cart
These disposable coveralls are designed with fabric and seams that have been rigorously tested for protection against blood and blood-borne pathogens.
  • Both the fabric and seams pass ASTM F1670 and F1671 tests for protection against blood, bloodborne pathogens, and bodily fluids
  • Durable fabric is built to withstand tears and abrasions in tough working environments
  • Zipper front with a sealable storm flap helps keep contaminants from entering or seeping through exposed zipper seams
  • Red fabric pull tab on the zipper makes it quick and easy to find so you can suit up or down in no time
  • Taped seams help keep contaminants out for added protection, while elastic wrists and ankles keep coveralls secure and in place
  • Convenient thumb and finger loops help keep sleeves from riding up while you work
  • Double-sided tape around wrists helps keeps gloves in place during donning
  • Storm flap over zipper helps prevent contaminants from entering through otherwise exposed zipper seams
  • Roomy one-piece raglan sleeves resist tears around shoulders
  • A gusseted crotch gives the user more freedom to squat, bend, and crouch without ripping fabric
  • Possible applications include emergency response, healthcare, category 3 black water, crime scene remediation, and animal research
Specifications
Product Type A50 Breathable Particle and Splash Protection Coveralls
Size Small
Gender Unisex
Material Multi-layer Barrier Laminate Fabric
Color White
Disposable Single-use
Waist Style Elastic at Back
Ankle Style Elastic
Cuff Style Elastic, Finger Loops
Neck Style Collared
Hood Type Standard
Closure Type 2-way Zipper with Sealed Storm Flap
Seam Type Serged, Taped
Sterility Non-sterile
Resistance Bloodborne Pathogens, Liquid, Particle, Tears
Latex Free Yes
No. of Pockets None