Learn More
Perkin Elmer US LLC White Calibration Material - 1.25 in
Catalog No. 502113277
Encompass_Preferred
PerkinElmer's calibrated diffuse reflectance standards sets are used to establish a spectrometer's photometric accuracy and linearity in diffuse reflectance. Reflectance calibration is NIST traceable. Calibrated 250-2500 nm reported each 50 nm. Sets include a disc with data for each 1 nm.
Catalog No. 50-211-3277 Supplier Perkin Elmer US LLC Supplier No. PELA9057
PerkinElmer's calibrated diffuse reflectance standards sets are used to establish a spectrometer's photometric accuracy and linearity in diffuse reflectance. Reflectance calibration is NIST traceable. Calibrated 250-2500 nm reported each 50 nm. Sets include a disc with data for each 1 nm.